Stampa…
Dibutyltin oxide, Msynth®plus
Fornitore: Merck
Attenzione
M-Clarity™ quality level = MQ400
8.45035.0250EA 227 CHF
8.45035.0250 8.45035.1000
Dibutyltin oxide, Msynth®plus
Dibutyltin oxide
Formula: C₈H₁₈OSn
Peso molecolare: 248,94 g/mol
Punto di fusione: > 300 °C
Densità: 1,6 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00001992
CAS Number: 818-08-6
EINECS: 212-449-1
UN: 3077
ADR: 9,III
Visualizza i codici
Solubility 0.004 g/l (20 °C)
Molar Mass 248.94 g/mol
Bulk Density 650 - 750 kg/m3
Boiling Point 300 °C decomposes
Density 1.5 g/cm3 (20 °C)
Ignition Point 525 °C
Hazard Class 9
Hazard Class Description Miscellaneous hazardous material
Learn more
About VWR
Mehr About VWR