Stampa…

2-Amino-3-hydroxypyridin 98%

Fornitore: BLD PHARMATECH GMBH

BLDPBD9602-100GEA 24 CHF
BLDPBD9602-100G BLDPBD9602-25G BLDPBD9602-500G
2-Amino-3-hydroxypyridin 98%
2-Amino-3-hydroxypyridin

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₅H₆N₂O
Peso molecolare: 110,12 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006317
CAS Number: 16867-03-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR