Stampa…

3,4-Dimethoxythiophenol 98%

BLDPBD82095-5GEA 72 CHF
BLDPBD82095-5G BLDPBD82095-25G BLDPBD82095-1G
3,4-Dimethoxythiophenol 98%
3,4-Dimethoxythiophenol

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₀O₂S
Peso molecolare: 170,23 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00052823
CAS Number: 700-96-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR