Stampa…

2-Fenossietanolo 94%

Synonyms: Fenil glicol

BLDPBD78011-100GEA 9 CHF
BLDPBD78011-100G BLDPBD78011-500G BLDPBD78011-1000G
2-Fenossietanolo 94%
2-Fenossietanolo

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₅OCH₂CH₂OH
Peso molecolare: 138,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002857
CAS Number: 122-99-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR