Stampa…

1-Butanesulphonic acid sodium salt 98%

Fornitore: BLD PHARMATECH GMBH

BLDPBD57587-10GEA 23 CHF
BLDPBD57587-10G BLDPBD57587-25G BLDPBD57587-5G
1-Butanesulphonic acid sodium salt 98%
1-Butanesulphonic acid sodium salt

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₉NaO₃S
Peso molecolare: 160,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007540
CAS Number: 2386-54-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR