Stampa…

2-Nitroresorcinol 99%

Fornitore: BLD PHARMATECH GMBH

BLDPBD3433-100GEA 141 CHF
BLDPBD3433-100G BLDPBD3433-25G BLDPBD3433-5G
2-Nitroresorcinol 99%
2-Nitroresorcinol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₆H₅NO₄
Peso molecolare: 155,11 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007124
CAS Number: 601-89-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR