Stampa…

2-Pyridinethiol 98%

Fornitore: BLD PHARMATECH GMBH

BLDPBD32947-500GEA 172 CHF
BLDPBD32947-500G BLDPBD32947-100G BLDPBD32947-25G BLDPBD32947-5G BLDPBD32947-0.5G
2-Pyridinethiol 98%
Pyridine-2(1H)-thione

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₅NS
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00457440
CAS Number: 2637-34-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR