Stampa…

2,3,5,6-Tetramethylpyrazine 95%

Fornitore: BLD PHARMATECH GMBH

BLDPBD30227-500GEA 109 CHF
BLDPBD30227-500G BLDPBD30227-0.5G BLDPBD30227-5G BLDPBD30227-25G BLDPBD30227-100G BLDPBD30227-10G
2,3,5,6-Tetramethylpyrazine 95%
2,3,5,6-Tetramethylpyrazine

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₁₂N₂
Peso molecolare: 136,2 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006146
CAS Number: 1124-11-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR