Stampa…

1,2,3-Benzotriazolo 98%

Fornitore: BLD PHARMATECH GMBH

BLDPBD29933-500GEA 16 CHF
BLDPBD29933-500G BLDPBD29933-1000G BLDPBD29933-100G
1,2,3-Benzotriazolo 98%
1,2,3-Benzotriazolo

Storage condition: Sealed in dry, Room Temperature

Formula: C₆H₅N₃
Peso molecolare: 119,13 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005699
CAS Number: 95-14-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR