Stampa…

Ferro (II) solfato eptaidrato 99+%

Fornitore: BLD PHARMATECH GMBH

BLDPBD219903-500GEA 13 CHF
BLDPBD219903-500G BLDPBD219903-1000G BLDPBD219903-25G BLDPBD219903-100G
Ferro (II) solfato eptaidrato 99+%
Ferro (II) solfato eptaidrato

Storage condition: Sealed in dry, Room Temperature

Formula: FeSO₄·7H₂O
Peso molecolare: 278,02 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149719
CAS Number: 7782-63-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR