Stampa…

Trifenil fosfato 99%

Fornitore: BLD PHARMATECH GMBH

BLDPBD155670-25GEA 7 CHF
BLDPBD155670-25G
Trifenil fosfato 99%
Trifenil fosfato

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₁₅O₄P
Peso molecolare: 326,29 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003031
CAS Number: 115-86-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR