Stampa…

Pirene 97%

BLDPBD149655-5GEA 6 CHF
BLDPBD149655-5G BLDPBD149655-10G BLDPBD149655-25G
Pirene 97%
Pirene

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₆H₁₀
Peso molecolare: 202,26 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00004136
CAS Number: 129-00-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR