Stampa…

Phenazine 97%

Fornitore: BLD PHARMATECH GMBH

BLDPBD14396-5GEA 7 CHF
BLDPBD14396-5G BLDPBD14396-25G BLDPBD14396-1G BLDPBD14396-500G BLDPBD14396-10G BLDPBD14396-100G
Phenazine 97%
Phenazine

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₂H₈N₂
Peso molecolare: 180,21 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00005023
CAS Number: 92-82-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR