Stampa…

Ethyl methanesulphonate 97%

BLDPBD129410-500GEA 426 CHF
BLDPBD129410-500G BLDPBD129410-5G BLDPBD129410-25G BLDPBD129410-100G BLDPBD129410-1G BLDPBD129410-10G
Ethyl methanesulphonate 97%
Ethyl methanesulphonate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₃H₈O₃S
Peso molecolare: 124,16 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00007559
CAS Number: 62-50-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR