Stampa…

Methanesulphonic anhydride 97%

Fornitore: BLD PHARMATECH GMBH

BLDPBD105771-25GEA 9 CHF
BLDPBD105771-25G BLDPBD105771-5G BLDPBD105771-10G
Methanesulphonic anhydride 97%
Methanesulphonic anhydride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂H₆O₅S₂
Peso molecolare: 174,2 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00007556
CAS Number: 7143-01-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR