Stampa…

2,3,5-Trimethylpyrazine 97%

Fornitore: BLD PHARMATECH GMBH

BLDPBD10043-25GEA 10 CHF
BLDPBD10043-25G BLDPBD10043-10G
2,3,5-Trimethylpyrazine 97%
2,3,5-Trimethylpyrazine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₇H₁₀N₂
Peso molecolare: 122,17 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006145
CAS Number: 14667-55-1

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR