Bismuth(III) fluoride 99.999%
Fornitore: Apollo Scientific
|
Pericolo
|
APOSPC7821-25GEA
475
CHF
APOSPC7821-25G
Bismuth(III) fluoride 99.999%
Bismuth(III) fluoride
Formula:
BiF₃ Peso molecolare: 265,98 g/mol Punto di fusione: 727 °C Densità: 5,32 g/cm³ (25 °C) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00010893 CAS Number: 7787-61-3 UN: 3260 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...