Stampa…

3-Bromobenzylamine 98%

Fornitore: Apollo Scientific
Pericolo

APOSOR40377-100GEA 416 CHF
APOSOR40377-100G APOSOR40377-25G
3-Bromobenzylamine 98%
3-Bromobenzylamine
Formula: C₇H₈BrN
Peso molecolare: 186,05 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01026119
CAS Number: 10269-01-9
UN: 2735
ADR: 8,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR