2,3,5-Trimethylpyrazine
Fornitore: Apollo Scientific
APOSOR350380-100GEA
103
CHF
APOSOR350380-100G
2,3,5-Trimethylpyrazine
2,3,5-Trimethylpyrazine
Formula:
C₇H₁₀N₂ Peso molecolare: 122,17 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00006145 CAS Number: 14667-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...