Stampa…

2,3-Dibromo-6-chloropyridine 95+%

APOSOR345145-10GEA 0 CHF
APOSOR345145-10G APOSOR345145-5G APOSOR345145-1G
2,3-Dibromo-6-chloropyridine 95+%
2,3-Dibromo-6-chloropyridine
Formula: C₅H₄ClN
Peso molecolare: 113,55 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06798229
CAS Number: 885952-16-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR