Stampa…

4-Chloro-2-hydroxypyridine

Fornitore: Apollo Scientific

APOSOR322384-1GEA 64.6 CHF
APOSOR322384-1G
4-Chloro-2-hydroxypyridine
4-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
Peso molecolare: 129,55 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03093022
CAS Number: 40673-25-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR