Stampa…

4-Bromophenyl isocyanate

Fornitore: Apollo Scientific

APOSOR322000-1GEA 122 CHF
APOSOR322000-1G
4-Bromophenyl isocyanate
4-Bromophenyl isocyanate

  • May form small amount of sediment.

Formula: C₇H₄BrNO
Peso molecolare: 198,02 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002022
CAS Number: 2493-02-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR