Stampa…

Tetrabutylphosphonium bromide

Pericolo

APOSOR315490-100GEA 0 CHF
APOSOR315490-100G APOSOR315490-500G
Tetrabutylphosphonium bromide
Tetrabutylphosphonium bromide
Formula: C₁₆H₃₆BrP
Peso molecolare: 339,34 g/mol
Punto di fusione: 104 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011853
CAS Number: 3115-68-2
EINECS: 211-487-8
UN: 3464
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR