Stampa…

2-Bromo-4-chloropyridine 97%

Fornitore: Apollo Scientific

APOSOR15630-25GEA 131 CHF
APOSOR15630-25G
2-Bromo-4-chloropyridine 97%
2-Bromo-4-chloropyridine
Formula: C₅H₃BrClN
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD03840757
CAS Number: 22918-01-0

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR