Stampa…

Thulium(III) fluoride, anidro 99,9%

APOSIN3604-25GEA 0 CHF
APOSIN3604-25G APOSIN3604-5G
Thulium(III) fluoride, anidro 99,9%
Thulium(III) fluoride
Formula: F₃Tm
Peso molecolare: 225,93 g/mol
Punto di fusione: 1158 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00049611
CAS Number: 13760-79-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR