Neodymium tribromide hydrate 99,9%
Fornitore: Apollo Scientific
|
Attenzione
|
APOSIN2626-25GEA
165
CHF
APOSIN2626-25G
APOSIN2626-100G
Neodymium tribromide hydrate 99,9%
Neodymium tribromide hydrate
Formula:
Br₃Nd·H₂O Peso molecolare: 401,97 g/mol Punto di fusione: 684 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00748137 CAS Number: 29843-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...