Stampa…

Holmium trinitrate pentahydrate 99,9%

Pericolo

APOSIN1987-50GEA 0 CHF
APOSIN1987-50G APOSIN1987-10G
Holmium trinitrate pentahydrate 99,9%
Holmium trinitrate pentahydrate
Formula: HoN₃O₉·5H₂O
Peso molecolare: 441,02 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149737
CAS Number: 14483-18-2
UN: 1477
ADR: 5.1,II

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR