Stampa…

Gadolinium trifluoride, anidro 99,9%

Fornitore: Apollo Scientific
Attenzione

APOSIN1828-25GEA 0 CHF
APOSIN1828-25G
Gadolinium trifluoride, anidro 99,9%
Gadolinium trifluoride
Formula: F₃Gd
Peso molecolare: 214,25 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00016113
CAS Number: 13765-26-9
EINECS: 237-369-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR