Stampa…

D-(-)-Arginine

Fornitore: Apollo Scientific
Attenzione

APOSBIA0864-1GEA 0 CHF
APOSBIA0864-1G APOSBIA0864-5G
D-(-)-Arginine
D-(-)-Arginine
Formula: C₆H₁₄N₄O₂
Peso molecolare: 174,2 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063116
CAS Number: 157-06-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR