Stampa…

5-Chloro-6-fluoroisatin

Fornitore: Apollo Scientific

APOSPC510278-1GEA 872 CHF
APOSPC510278-1G
5-Chloro-6-fluoroisatin
5-Chloro-6-fluoroisatin
Formula: C₈H₃ClFNO₂
Peso molecolare: 199,566 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD09728573
CAS Number: 953897-06-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR