Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Fornitore: Apollo Scientific
APOSPC510265-1GEA
1470
CHF
APOSPC510265-1G
Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Ethyl 3-[Hydroxy[4-(trifluoromethyl)phenyl]methyl]isoxazole-4-carboxylate
Formula:
C₁₄H₁₂F₃NO₄ Peso molecolare: 315,245 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28977400 CAS Number: 2006278-06-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...