Stampa…

3-(3-Chlorophenyl)-3-fluoropyrrolidine

Fornitore: Apollo Scientific

APOSPC510218-1GEA 1470 CHF
APOSPC510218-1G
3-(3-Chlorophenyl)-3-fluoropyrrolidine
3-(3-Chlorophenyl)-3-fluoropyrrolidine
Formula: C₁₀H₁₁ClFN
Peso molecolare: 199,652 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD26038927
CAS Number: 1565489-33-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR