Stampa…

Methyl 4-(Trifluoromethyl)benzimidate hydrochloride

Fornitore: Apollo Scientific

APOSPC510180-5GEA 477 CHF
APOSPC510180-5G
Methyl 4-(Trifluoromethyl)benzimidate hydrochloride
Methyl 4-(Trifluoromethyl)benzimidate hydrochloride
Formula: C₉H₈F₃NO·ClH
Peso molecolare: 239,622 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD12031909
CAS Number: 56108-08-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR