3,5-Difluoro-4-(methylsulphonyl)phenol
Fornitore: Apollo Scientific
APOSPC510174-1GEA
1350
CHF
APOSPC510174-1G
3,5-Difluoro-4-(methylsulphonyl)phenol
3,5-Difluoro-4-(methylsulphonyl)phenol
Formula:
C₇H₆F₂O₃S Peso molecolare: 208,183 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD27986812 CAS Number: 1447123-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...