4-(tert-Butyl)-3,5-difluorophenol
Fornitore: Apollo Scientific
APOSPC510173-1GEA
1110
CHF
APOSPC510173-1G
4-(tert-Butyl)-3,5-difluorophenol
4-(tert-Butyl)-3,5-difluorophenol
Formula:
C₁₀H₁₂F₂O Peso molecolare: 186,198 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD20485342 CAS Number: 910486-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...