2-Amino-4-fluoro-1-methylbenzimidazole
Fornitore: Apollo Scientific
APOSPC510163-1GEA
1100
CHF
APOSPC510163-1G
2-Amino-4-fluoro-1-methylbenzimidazole
2-Amino-4-fluoro-1-methylbenzimidazole
Formula:
C₈H₈FN₃ Peso molecolare: 165,168 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD12149741 CAS Number: 1183473-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...