Stampa…

Ethyl 3-Bromo-5-fluoroindole-1-carboxylate

Fornitore: Apollo Scientific

APOSPC510118-5GEA 855 CHF
APOSPC510118-5G
Ethyl 3-Bromo-5-fluoroindole-1-carboxylate
Ethyl 3-Bromo-5-fluoroindole-1-carboxylate
Formula: C₁₁H₉BrFNO₂
Peso molecolare: 286,097 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD22205880
CAS Number: 1375064-48-4

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR