Stampa…

Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate

Fornitore: Apollo Scientific

APOSPC510109-5GEA 1100 CHF
APOSPC510109-5G
Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Methyl 1-Cbz-3-fluoropiperidine-3-carboxylate
Formula: C₁₅H₁₈FNO₄
Peso molecolare: 295,306 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD21641308
CAS Number: 1363166-15-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR