Stampa…

4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid

Fornitore: Apollo Scientific

APOSPC510083-5GEA 855 CHF
APOSPC510083-5G
4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid
4-(4-Benzyl-1-piperazinyl)-3-fluorobenzoic acid
Formula: C₁₈H₁₉FN₂O₂
Peso molecolare: 314,354 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD20134209
CAS Number: 1346597-48-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR