2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
Fornitore: Apollo Scientific
APOSPC502877-1GEA
2110
CHF
APOSPC502877-1G
2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether 95%
2-Amino-5-iodophenyl 2,6-difluoro-4-(trifluoromethyl)phenyl ether
Formula:
C₁₃H₇F₅INO Peso molecolare: 415,097 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD31629954 CAS Number: 2244083-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...