Stampa…

Methyl 5-fluoro-2-methoxybenzoate

Fornitore: Apollo Scientific

APOSPC502593-1GEA 89 CHF
APOSPC502593-1G
Methyl 5-fluoro-2-methoxybenzoate
Methyl 5-fluoro-2-methoxybenzoate
Formula: C₉H₉FO₃
Peso molecolare: 184,164 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD06203734
CAS Number: 151793-20-3

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR