Methyl 3-fluoro-2-iodobenzoate 95+%
Fornitore: Apollo Scientific
APOSPC502586-1GEA
90.8
CHF
APOSPC502586-1G
Methyl 3-fluoro-2-iodobenzoate 95+%
Methyl 3-fluoro-2-iodobenzoate
Formula:
C₈H₆FIO₂ Peso molecolare: 280,035 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD18399305 CAS Number: 393-78-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...