Stampa…

2-Amino-5-fluoro-4-nitrophenol 97%

Fornitore: Apollo Scientific

APOSPC501727-250MGEA 294 CHF
APOSPC501727-250MG
2-Amino-5-fluoro-4-nitrophenol 97%
2-Amino-5-fluoro-4-nitrophenol
Formula: C₆H₅FN₂O₃
Peso molecolare: 172,114 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28967930
CAS Number: 1850312-96-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR