Stampa…

5-Bromo-2-fluoro-4-methylbenzoic acid

Fornitore: Apollo Scientific

APOSPC501554-1GEA 287 CHF
APOSPC501554-1G
5-Bromo-2-fluoro-4-methylbenzoic acid
5-Bromo-2-fluoro-4-methylbenzoic acid
Formula: C₈H₆BrFO₂
Peso molecolare: 233,034 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD23712067
CAS Number: 515135-65-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR