Stampa…

5-Fluoro-3-nitropicolinonitrile 97%

Fornitore: Apollo Scientific

APOSPC48748-250MGEA 87.2 CHF
APOSPC48748-250MG
5-Fluoro-3-nitropicolinonitrile 97%
5-Fluoro-3-nitropicolinonitrile
Formula: C₆H₂FN₃O₂
Peso molecolare: 167,097 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD11040666
CAS Number: 1033202-51-5

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR