6-Amino-3-chloro-2-fluorobenzoic acid 96%
Fornitore: Apollo Scientific
|
Pericolo
|
APOSPC450566-100MGEA
102
CHF
APOSPC450566-100MG
6-Amino-3-chloro-2-fluorobenzoic acid 96%
6-Amino-3-chloro-2-fluorobenzoic acid
Formula:
C₇H₅ClFNO₂ Peso molecolare: 189,572 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD03618452 CAS Number: 874782-54-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...