3,3,4,4,5,5,6,6,6-Nonafluorohexyl-1-ene carbonate
Fornitore: Apollo Scientific
APOSPC450491-250MGEA
765
CHF
APOSPC450491-250MG
3,3,4,4,5,5,6,6,6-Nonafluorohexyl-1-ene carbonate
3,3,4,4,5,5,6,6,6-Nonafluorohexyl-1-ene carbonate
Formula:
C₇H₃F₉O₃ Peso molecolare: 306,083 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD28901326 CAS Number: 1469980-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...