Stampa…

sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate

Fornitore: Apollo Scientific

APOSPC450476-25GEA 1840 CHF
APOSPC450476-25G
sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
sec-Butyl 2,2,3,4,4,4-hexafluorobutyl carbonate
Formula: C₉H₁₂F₆O₃
Peso molecolare: 282,18 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334448
CAS Number: 1980035-12-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR