Stampa…

tert-Butyl 1H,1H,7H-perfluorohexyl carbonate

Fornitore: Apollo Scientific

APOSPC450467-25GEA 1840 CHF
APOSPC450467-25G
tert-Butyl 1H,1H,7H-perfluorohexyl carbonate
tert-Butyl 1H,1H,7H-perfluorohexyl carbonate
Formula: C₁₂H₁₂F₁₂O₃
Peso molecolare: 432,203 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334440
CAS Number: 1980038-41-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR