Stampa…

Butyl 1H,1H,9H-perfluorononyl carbonate

Fornitore: Apollo Scientific

APOSPC450455-25GEA 1840 CHF
APOSPC450455-25G
Butyl 1H,1H,9H-perfluorononyl carbonate
Butyl 1H,1H,9H-perfluorononyl carbonate
Formula: C₁₄H₁₂F₁₆O₃
Peso molecolare: 532,218 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD28334429
CAS Number: 1980034-58-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR